Description
- Fill prescription at a local vision center and fit the inserts into the safety glasses
- Tend to be more lightweight and comfortable than options like side shields and over-the-glasses safety glasses
- Polycarbonate lenses help protect the prescription insert from damage and provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Wraparound frame has a base curve of 6.5 that extends the range of peripheral protection, as well as adjustable ratchet-action temples for with five positions for a customized fit and lens angle
- Corrective lenses are hard coated for scratch resistance and longer life
- Each pair comes with a neck cord for securing
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection
- Stylish, comfortable and functional for construction, manufacturing, and a host of other industries
Specifications
Specifications
| Lens Coating | Anti-fog |
| Certifications/Compliance | Meets ANSI Z87.1+ standards for high impact (38507-00 meets ANSI Z87.1+2010 and 38507-01 meets ANSI Z87.1+2015) |
| Lens Tint | Indoor/Outdoor |
| Product Type | Safety Eyewear |
| Frame Color | Metallic Blue |
| Lens Material | Polycarbonate |
| Frame Type | Metallic blue frame with black tips |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.