Learn More
Description
- Designed for entry-level protection from hazardous dust
- Feature a zipper front, attached hood, serged seams and elastic wrists
- Five layers of stacked barrier protection, for a lightweight, breathable protective coverall
- Ideal for food processing, general maintenance and manufacturing
Specifications
Specifications
| Product Type | General Purpose Coveralls |
| Size | X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Closure Type | Front Zipper |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.