Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A10 Coveralls

Catalog No. 19041843 Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-041-843 2X-Large
19-041-841 Large
19-041-844 3X-Large
19-041-842 X-Large
19-041-845 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-041-843 Supplier Ansell Supplier No. 12226
This item is not returnable. View return policy

A10 Coveralls provide economical, disposable protection for workers exposed to dirt and grime in the workplace.

  • Designed for entry-level protection from hazardous dust
  • Feature a zipper front, attached hood, serged seams and elastic wrists
  • Five layers of stacked barrier protection, for a lightweight, breathable protective coverall
  • Ideal for food processing, general maintenance and manufacturing
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ANSI ISEA 101-1996
Latex Free Yes
For Use With (Application) Food Processing, General Maintenance, Manufacturing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.