Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A10 Light Duty Beard Cover
GREENER_CHOICE

Catalog No. p-3202971
Click to view available options
Size:
Universal
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Ideal for food processing, general maintenance, and manufacturing

  • Comfortable
  • Lightweight polypropylene material
TRUSTED_SUSTAINABILITY

Specifications

Type General Purpose Beard Cover
Size Universal
Material Polypropylene
Color White
Disposable Single-use
For Use With (Application) Food Processing, General Maintenance, and Manufacturing
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.