Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Pants
Catalog No. 17981250 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-981-250 Medium
17-981-251 Large
17-981-252 X-Large
17-981-253 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-981-250 Supplier Ansell Supplier No. 36222
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Lightweight, abrasion-resistant particle protection pants
  • Breathable, patented Microforce™ barrier SMS fabric
  • Barrier protection against dry particulates
  • Strong and abrasion-resistant
  • Elastic waist; no pockets
Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments
Compliance
Meets NFPA 99 criteria for antistatic materials
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Gender Unisex
Material SMS
Color White
Disposable Single-use
Sterility Non-sterile
Waist Style Elastic