Description
- Breathable, patented Microforce™ barrier SMS fabric
- Barrier protection against dry particulates
- Strong and abrasion-resistant
- Elastic waist; no pockets
Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments
Compliance
Meets NFPA 99 criteria for antistatic materials
Specifications
Specifications
| Size | X-Large |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Disposable | Single-use |
| Certifications/Compliance | NFPA 99 Criteria for Antistatic Materials |
| Sterility | Non-sterile |
| Waist Style | Elastic |