Learn More
Ansell KleenGuard™ A20 Breathable Particle Protection Pants
Catalog No. 17981253 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-981-253 2X-Large
17-981-250 Medium
17-981-251 Large
17-981-252 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-981-253 Supplier Ansell Supplier No. 36225
This item is not returnable. View return policy
Lightweight, abrasion-resistant particle protection pants
  • Breathable, patented Microforce™ barrier SMS fabric
  • Barrier protection against dry particulates
  • Strong and abrasion-resistant
  • Elastic waist; no pockets
Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments
Compliance
Meets NFPA 99 criteria for antistatic materials
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Gender Unisex
Material SMS
Color White
Disposable Single-use
Certifications/Compliance NFPA 99 Criteria for Antistatic Materials
Sterility Non-sterile
Waist Style Elastic