Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Frocks
GREENER_CHOICE

Catalog No. 19169881 Shop All Ansell Products
Change view
Click to view available options
Size:
3X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-881 3X-Large
19-457-221 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-169-881 Supplier Ansell Supplier No. 30937
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Frocks feature breathable, patented MICROFORCE barrier SMS fabric and meets NFPA 99 criteria for antistatic materials.

  • Snap front closure with snap at collar
  • Long length
  • Elastic wrists
  • No pockets
  • Passes NFPA 99 criteria for antistatic materials
  • Typical uses: Laboratories; Decontamination modesty garment; Remediation; Pesticides; Heavy dust environments

Order Info

This size is available on a made-to-order basis – 30 case minimum order per product code; product available within 8 weeks of order receipt

TRUSTED_SUSTAINABILITY

Specifications

Size 3X-Large
Material SMS
Color White
Closure Type Snaps
Cuff Style Elastic
Certifications/Compliance NFPA 99
Product Type General Purpose Frock
For Use With (Application) Laboratories, Decontamination, Remediation, Pesticides, Heavy Dust Environments

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.