Learn More
Ansell KleenGuard™ Eyewear: V10
Offer comfort, flexibility, and protection
Specifications
Lens Coating Uncoated
UV Protection 0.999
Frame Type Lightweight
View More Specs
Products 1
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Price Quantity & Availability  
19-167-760
View Documents
Ansell
0865100
Amber N/A N/A N/A
Description
Description
  • Impact-resistant lenses offer 99.9% UVA/UVB protection
  • Amber: For low-light environments where contrast is low; blocks out blue light to highlight and define shapes
  • Smoke: For applications where eyestrain is a concern or bright, sunny days requiring maximum glare reduction
  • Indoor/Outdoor: For moving between indoor/shaded areas to direct sunlight and back
V10 Standard Model
  • Lightweight frames for added comfort
Compliance
Meet ANSI™ Z87.1-2003 standards.
Specifications
Specifications
Uncoated
0.999
Lightweight
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.