Learn More
Ansell Oasis™ Undergarment Coveralls
Catalog No. 19057456B Shop All Ansell Products
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-057-456B Large
19-057-456D 2X-Large
19-057-456C X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-057-456B Supplier Ansell Supplier No. 58403
This item is not returnable. View return policy
Keep cool while wearing hot protective apparel
Hot, nonbreathable protective apparel can become very uncomfortable as perspiration builds up after hours of wear. These coveralls, worn under protective apparel, are made of a lightweight, three-layer fabric that works with the skin's natural evaporation action to wick perspiration away from the skin. This, plus the fabric's quick-drying capability, greatly increases comfort. Cut in the Reflex™ coverall design, with roomier fit in key stress areas for greater mobility and longer wear. Front zipper, elastic in the back, wrists and ankles. Disposable, eliminating inconvenience and cost of laundering. Blue.
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size Large
Gender Unisex
Color Blue
Disposable Single-use
Neck Style Collared
Sterility Non-sterile