Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell PFR95™ N95 Particulate Filter Respirators

Catalog No. 189991444 Shop All Ansell Products
Change view
Click to view available options
Color:
Orange
White
Quantity:
35/Pk.
50/Pk.
Size:
Regular
Small
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Quantity Size
18-999-1444 Orange 35/Pk. Small
18-999-1445 Orange 35/Pk. Regular
18-999-4812 White 50/Pk. Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-999-1444 Supplier Ansell Supplier No. 46827

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Particulate filter respirator (PFR) features a pouch-style design to provide a large breathing area while limiting facial contact.

  • Bendable aluminum nosepiece for comfortable, secure fit
  • Large breathing chamber
  • Two polyurethane headbands secure the mask to help ensure a tight facial seal and prevent leakage
  • Bacterial filtration efficiency: 95% at 0.3 μm
  • Patented construction that provides protection from bloodborne pathogens and body fluids
TRUSTED_SUSTAINABILITY

Specifications

Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color Code Orange
Color Orange
Design Type Pouch/Duckbill
Disposable Single-use
Filter Type Surgical N95
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Small
With Exhalation Valve No
Filter Efficiency 95%
For Use With (Application) Provides protection from blood borne pathogens and body fluids
Material Headstrap Polyurethane
Model 46827
Nose Piece Material Aluminum
Nosepiece Type Adjustable Nose Clip
Quantity 35/Pk.
Resistant To Fluid
Strap Attachment Type Dual Headbands
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.