Learn More
Ansell PFR95™ N95 Particulate Filter Respirators
Particulate filter respirator (PFR) features a pouch-style design to provide a large breathing area while limiting facial contact.
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Design Type Pouch/Duckbill
Disposable Single-use
Filter Type Surgical N95
View More Specs
Products 3
Catalog Number Mfr. No. Color Quantity Size Price Quantity & Availability  
Catalog Number Mfr. No. Color Quantity Size Price Quantity & Availability  
18-999-1444
View Documents
Ansell
46827
Orange 35/Pk. Small N/A N/A N/A
18-999-1445
View Documents
Ansell
46727
Orange 35/Pk. Regular N/A N/A N/A
18-999-4812
View Documents
Ansell
62126
White 50/Pk. Regular N/A N/A N/A
Description
Description
Particulate filter respirator (PFR) features a pouch-style design to provide a large breathing area while limiting facial contact.
  • Bendable aluminum nosepiece for comfortable, secure fit
  • Large breathing chamber
  • Two polyurethane headbands secure the mask to help ensure a tight facial seal and prevent leakage
  • Bacterial filtration efficiency: 95% at 0.3 μm
  • Patented construction that provides protection from bloodborne pathogens and body fluids
Specifications
Specifications
N - Not Resistant to Oil
Pouch/Duckbill
Surgical N95
No
Provides protection from blood borne pathogens and body fluids
Aluminum
Fluid
No
NIOSH 42 CFR 84, N95
Single-use
Healthcare Particulate Respirator and Surgical Mask
95%
Polyurethane
Adjustable Nose Clip
Dual Headbands
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.