Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves

Catalog No. 19168879 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-168-879 Medium
19-168-878 Small
19-168-880 Large
19-168-881 X-Large
19-168-882 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-168-879 Supplier KleenGuard™ Supplier No. 13838
Add to Cart
Add to Cart
This item is not returnable. View return policy

Multi-purpose protective gloves designed for tasks that require high dexterity and fine motor skills. The thin-mil design provides tactile sensitivity, making KleenGuard™ G40 Black Polyurethane Coated Nylon Gloves ideal for performing delicate tasks such as electronics assembly.

  • General purpose protective gloves designed for tasks that require high dexterity and fine motor skills
  • Thin mil design provides tactile sensitivity, ideal for performing delicate tasks
  • Seamless knit nylon backing enhances durability, and provides resistance to tears and abrasion
  • Dark color is ideal for hiding dirt
TRUSTED_SUSTAINABILITY

Specifications

Coated Area Palm, Fingers
Coating Material Polyurethane
Color Black
Exterior Finish Partially Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Medium
Style Full-hand Mitt
For Use With (Application) Small Parts Handling, Precision Assembly, and Inspection
Glove Cuff Style Knit
Series G40
Length (English) 9.1 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.