Learn More
Ansell KleenGuard™ V80 MRXV™ Safety Goggles
Catalog No. 1798683B Shop All Ansell Products
Change view
Click to view available options
Description:
Standard lens
VisiClear™ Anti-fog
Frame Color:
Blue
Gray
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Frame Color
17-986-83B Standard lens Gray
17-986-83D VisiClear™ Anti-fog Blue
17-986-83E VisiClear™ Anti-fog Gray
17-986-83L Standard lens Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-986-83B Supplier Ansell Supplier No. 16678
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
KleenGuard V80 MXRV Safety Goggles give you and your workers reliable splash protection in an unventilated design. These splash goggles have a simple, sleek design and a curved lens for an enhanced vision field, while being roomy enough to fit over most prescription eye glasses.
  • Clear, optical-grade polycarbonate lens absorbs 99.9% of UV radiation up to 380nm
  • Oversized PVC frame for a snug, comfortable fit
  • Dual ventilation ports for unobstructed peripheral vision
  • Chemical-resistant neoprene headband attaches to ports on the goggle frame for easy adjustment
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Uncoated
Certifications/Compliance ANSI Z87.1-2010
Description Standard lens
Lens Tint Clear
Product Type Safety goggles
Frame Color Gray
Material Polycarbonate
Frame Material PVC