Learn More
Ansell KleenGuard™ V50 Cudas™ Safety Eyewear
Catalog No. 18052 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Clear
Smoke
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
18-052 Smoke
18-051 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-052 Supplier Ansell Supplier No. 14467
This item is not returnable. View return policy
KleenGuard™ V50 Cudas Safety Eyewear have a nylon frame designed for smaller, slimmer heads.
  • Wraparound optical-grade polycarbonate lens with integral sideshields offer 170° of distortion-free peripheral vision
  • 99.9% UV protection up to 380nm
  • Sleek design features built-in brow for protection and separate nylon nosepiece for snug, nonslip fit
  • Available with uncoated clear lens or uncoated smoke tint to reduce eyestrain in sunny, outdoor environments
Compliance
Meet ANSI™ Z87.1-2010 standards
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Meets ANSI Z87.1-2003standards; SEI certified.
Lens Tint Smoke
Lens Material Polycarbonate
Frame Material Nylon