Learn More
Ansell KleenGuard™ V50 Cudas™ Safety Eyewear
KleenGuard™ V50 Cudas Safety Eyewear have a nylon frame designed for smaller, slimmer heads.
Supplier: Ansell 14464
This item is not returnable.
View return policy
Description
- One size
- Curved lens
- Nylon frame
- Cudaside adjustable temples
- 99.9% UVA/UVB/UVC protection
- Meets ANSI Z87.1+ standards
Compliance
Meet ANSI™ Z87.1-2010 standards
Specifications
Meets ANSI Z87.1-2003standards; SEI certified. | |
CudasSlide | |
Nylon |
Clear | |
Polycarbonate |