Description
- Soft, comfortable self-adjusting rubber nose pad
- Precise placement of lens inserts
- 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Certifications/Compliance | ANSI Z87.1-2010 |
| Lens Tint | Yellow |
| Product Type | Safety Readers |
| Frame Color | Black |
| Magnification Power | +2.50 |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |
| UV Protection | 0.999 |