Learn More
Ansell KleenGuard™ V60 30-06™ Readers Safety Eyewear
KleenGuard V60 30-06 Readers Safety Glasses are equipped to suit those who need vision assistance and eye protection. Available in a variety of lens and frame colors and prescription strengths, they are designed for long-term comfort.
Supplier:  Ansell 19875
 View more versions of this product
Catalog No. 19-170-005
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
  • Soft, comfortable self-adjusting rubber nose pad
  • Precise placement of lens inserts
  • 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANSI Z87.1-2010
Safety Readers
+2.50
Nylon
Brown
Tortoise Shell
Polycarbonate
UV400
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.