Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Scott™ Control Antimicrobial Foam Skin Cleanser (0.1% Benzalkonium Chloride)

Catalog No. 19413007 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Quantity:
6 Pk.
Product Type:
Anti-bacterial Foam Soap
E-2 Foam Skin Cleanser
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
19-413-007 6 Pk. Anti-bacterial Foam Soap
19-413-008 6 Pk. E-2 Foam Skin Cleanser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-413-007 Supplier Kimberly-Clark Professional™ Supplier No. 91554
Only null left
Add to Cart
Add to Cart

Contains (0.1% Benzalkonium Chloride) to kill harmful germs such as E. Coli, Listeria, and Salmonella. Contains aloe and glycerin to moisturize the skin. Does not contain triclosan. High capacity for less refilling and maintenance time. Easy-to-load cassette clicks right into place. Bottle is recyclable after the pump is removed.

  • 6 cassettes / case; 1.0 L each; Scott Control E-2 Antimicrobial Foam Skin Cleanser contains 0.1% Benzalkonium Chloride and kills 99.9% of the most germs in as little as 15 seconds
  • It kills germs including Salmonella, Staphylococcus aureaus, and E. coli
  • This foaming commercial hand soap clear (dye-free), has a pleasant fruit scent
  • Easy-to-load, high capacity cassette clicks right into place and is compatible with the Scott Control Professional Cassette Skin Care Dispenser
  • The bottle is recyclable after the pump is removed

Specifications

Product Type Anti-bacterial Foam Soap
Color Clear
Composition Triclosan 0.5%, Aloe and Provitamin B5
For Use With (Application) To kill harmful germs such as E. Coli, Staphylococcus aureaus and Salmonella
For Use With (Equipment) Kimberly-Clark Professional™ Cassette skin care dispenser
Packaging Dispenser
Quantity 6 Pk.
Volume (English) 0.26 gal.
Volume (Metric) 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.