Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ A5 Cleanroom Apparel, Bulk Packaging
BIOPHARMAGREENER_CHOICE

Catalog No. 19169057 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large, 7X-Large, 8X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-057 3X-Large
19-169-052 Small
19-169-053 Medium
19-169-054 Large
19-169-055 X-Large
19-169-056 2X-Large
19-169-058 4X-Large
19-169-059 5X-Large
19-169-060 6X-Large, 7X-Large, 8X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-169-057 Supplier Ansell Supplier No. 49836
Add to Cart
Add to Cart
This item is not returnable. View return policy

Coveralls feature a polypropylene breathable SMS fabric with a cloth-like feel.

Kimtech™ A5 Cleanroom Coveralls are made of 100% polypropylene breathable SMS fabric, which gives the wearer cloth-like comfort. While other coveralls for cleanroom environments may look similar, the ones from Kimtech™ Pure have distinct advantages including tightly bound seams to strengthen the barrier, a high neck collar to reduce contamination, thumb loops to keep the sleeves from rolling back and a zipper flap to cover the zipper area.

In addition, this protective apparel is recyclable through the RightCycle program.

This personal protective apparel is triple-bagged and vacuum packaged, suitable for ISO Class 5 or higher cleanrooms. It is not made with natural rubber latex, silicone, or the BHT preservative and is designed for single-use (disposable PPE).

For use in biotech, medical device, electronics and telecommunications manufacturing, they are designed to protect your process by providing comfortable, sustainable protection.

  • Made of breathable SMS fabric, the coveralls feature a high-neck collar, a zipper flap, thumb loops and bound seams
  • Triple bagged, vacuum packed, suitable for ISO Class 5 or higher cleanrooms; CE certified
  • Kimtech™ A5 Cleanroom Apparel is manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
  • Recyclable through the RightCycle program, which can help you meet your sustainability goals
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Material Polypropylene, SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
Sterility Non-sterile
Collar Type Standing
For Use With (Application) Compounding, Mixing, Filling, Cleaning, Pharmaceutical, Biomedial Research
Cleanroom Class Class 100-100,000 (ISO 5-8)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.