Learn More
Description
- Available in bulk cleanroom packaging (double bag)
- Anti-static treatment and are low lint to reduce contamination risk
- High collar for increased neck protection
- Comfortable thumb loops to maintain glove and garment interface
- Disposable lab coats are recyclable through the RightCycle Program
- Enhanced chemical and biological protection (passes ASTM F1670 for penetration of blood and body fluids)
- Protection for people and processes against liquid splashes
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Size | X-Large |
| Material | Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Static |
| Collar Type | High Mandarin Collar |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

