Learn More
Ansell Kimtech™ Pure G3 EvT Nitrile Gloves
Catalog No. 19169455 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-455 2X-Large
19-169-450 X-Small
19-169-451 Small
19-169-452 Medium
19-169-453 Large
19-169-454 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-169-455 Supplier Ansell Supplier No. 38705
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Non-sterile, nitrile gloves designed to have low particles and extractables and are cleaned to meet the needs of a ISO Class 3 or higher critical environment.
  • Natural rubber latex-free reduces the risk of Type I allergic reactions
  • Recommended for ISO Class 3 and higher environments
  • Low particles and extractables
  • Cleaned to meet the needs of the critical cleanroom environment
  • Textured and tacky surface provides secure grip
  • AQL 1.5 for pinholes
  • Ambidextrous
  • Ideal for critical tasks in semiconductor, electronics and medical device manufacturing facilities
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D5151
Cleanroom Class Class 1 (ISO 3)
AQL 1.5
Color White
Exterior Finish Fully Textured
Finger Thickness (English) 5 mil
Finger Thickness (Metric) 0.12 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Size 2X-Large
Sterility Non-sterile
Thickness (English) 5 mil
Thickness (Metric) 0.12 mm
Model 38705
Series Kimtech™ Pure G3 EvT
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.