Learn More
Kimtech™ Pure G5 Sterile Latex Gloves
Catalog No. 19048586G Shop All Ansell Products
Change view
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
10
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-048-586G 9
19-048-586A 6
19-048-586B 6.5
19-048-586C 7
19-048-586D 7.5
19-048-586E 8
19-048-586F 8.5
19-048-586H 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-048-586G Supplier Kimtech™ Supplier No. 56857
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Sterile, hand-specific latex gloves provide sensitive and dextrous protection in Class 100/ISO 5 or higher cleanroom environments.
  • Fully sterilized latex material provides high levels of contamination protection
  • Hand-specific, disposable and a natural color
  • Textured palm and fingertips enhance grip and tactile sensitivity
  • Beaded cuffs add strength to the gloves reducing the risk of tearing
  • Easy donning and doffing
  • Made from latex sterilized with gamma radiation and containing no powder
  • Suitable for use in EU GMP ISO Class 5 Grade A or higher sterile cleanrooms
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM D3577, IEST-RP-CC005
Certifications/Compliance (Europe) PPE Cat III, EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016
Cleanroom Class Class 100 (ISO 5)
AQL 1.5
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Specific
Material Latex
Powdered Powder Free
Size 9
Sterility Sterile
Thickness (English) 8 mil
Thickness (Metric) 0.20 mm
Model 56857
Series Kimtech™ Pure G5
Length (English) 12 in.
Length (Metric) 30.5 cm