Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A10 Light-Duty Coveralls

Catalog No. 19812875 Shop All Ansell Products
Change view
Click to view available options
Description:
With Attached Hood and Boots
Without Hood and Boots
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-812-875 Without Hood and Boots Large
19-812-877 Without Hood and Boots 2X-Large
19-812-881 With Attached Hood and Boots X-Large
19-812-880 With Attached Hood and Boots Large
19-812-883 With Attached Hood and Boots 3X-Large
19-812-878 Without Hood and Boots 3X-Large
19-812-882 With Attached Hood and Boots 2X-Large
19-812-876 Without Hood and Boots X-Large
19-812-879 Without Hood and Boots 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-812-875 Supplier Ansell Supplier No. 10468
This item is not returnable. View return policy

Light-duty coveralls are economically priced for protection in nonhazardous environments.

  • Lightweight, breathable fabric keeps workers cool and comfortable
  • Zipper front
  • Elastic wrists
  • Available with or without attached hood and boots
  • White

Compliance

Meets ANSI/ISEA 101-1996 standards.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Light-duty Coveralls
Size Large
Gender Unisex
Material Polypropylene
Color White
Disposable Limited-use
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Certifications/Compliance ANSI/ISEA 101-1996
Description Without Hood and Boots
Model KleenGuard™ A10
For Use With (Application) Food Processing, General Maintenance, Manufacturing
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.