Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Coveralls, Open Wrists and Ankles
GREENER_CHOICE

Catalog No. 1798144C Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-981-44C 2X-Large
17-981-44E 4X-Large
19-047-770 Medium
17-981-44A Large
17-981-44D 3X-Large
17-981-44B X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Not for use near heat, flames or flammable environments
Catalog No. 17-981-44C Supplier Ansell Supplier No. 49005
Add to Cart
Add to Cart
This item is not returnable. View return policy
Not for use near heat, flames or flammable environments

Coveralls are designed to protect against particles such as dirt, grime, and dust. Made with breathable SMS fabric with five layers of stacked barrier protection.

  • For use in laboratories, remediation, decontamination, pesticides, and heavy dust environments.
  • Open wrists and ankles, zippered front.
  • Provides general protection against many particles
  • Breathable patented Microforce™ barrier SMS fabric passes NFPA 99 for antistatic materials.
  • High abrasion resistance, low linting
  • Withstands multiple wearings
  • Air and moisture vapor can pass through to aid wearer comfort.
  • Triple-stitched serged seams.
  • Offer particle holdout coupled with unmatched comfort attributes.
  • Fabric is comprised of five layers of stacked barrier protection (SMMMS).
  • Human-centric Reflex™ design offers comfortable fit and freedom of movement.
  • Eligible for recycling via RightCycle™ when used in non-hazardous applications as classified by federal, state, and local regulations.
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Open
Cuff Style Open
Closure Type Front Zipper
Sterility Non-sterile
Resistance Dust
Certifications/Compliance NFPA 99
Description Microforce™ Barrier SMS Fabric
Model KleenGuard™ A20
For Use With (Application) Laboratories, Decontamination Modesty Garment, Remediation, Pesticides, Heavy Dust Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.