Learn More
Description
A20 lab coats are comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
- Tailored style made of breathable patented MICROFORCE™ Barrier SMS Fabric
- Knee-length, serged seams
- Long sleeves, notched-lapel collar
- Air and moisture vapor can pass through to aid wearer comfort
- Meets NFPA 99 criteria for antistatic materials
- Two styles to choose from:
- Open wrists with pockets, snap closure
- Elastic wrists without pockets, hook and loop closure
Compliance
NFPA 99 compliant antistatic material
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | 2X-Large |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Open |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Style | Knee-length |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

