Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A30 Coveralls, Open Wrists and Ankles
GREENER_CHOICE

Catalog No. 19170386A Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-170-386A Medium
19-170-386E 3X-Large
19-170-386B Large
19-170-386D 2X-Large
19-170-386C X-Large
19-170-386F 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-170-386A Supplier Ansell Supplier No. 46002
Add to Cart
Add to Cart
This item is not returnable. View return policy

Limited-use coveralls designed to protect against hazardous dry particles, aerosols and low-hazardous limited liquid splash.

  • Air permeable SMS fabric incorporates a web of microfibers that filter particulates and water-based liquids
  • Suitable for many applications including pharmaceutical research and food processing
  • Provides protection from low-hazardous particles and liquids
  • Strong and abrasion-resistant
  • Suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5 cm) flap
  • Open wrists and ankles
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Open
Cuff Style Open
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Sterility Non-sterile
Resistance Particle, Liquid
Back Style Elastic
Certifications/Compliance NFPA 99
Model KleenGuard™ A30
For Use With (Application) Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.