Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A30 Coveralls with Elastic Back, Wrists, Ankles, Attached Hood
GREENER_CHOICE

Catalog No. 19170388D
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Limited-use coveralls designed to protect against hazardous dry particles, aerosols and low-hazardous limited liquid splash.

  • Air permeable SMS fabric incorporates a web of microfibers that filter particulates and water-based liquids
  • Suitable for many applications including pharmaceutical research and food processing
  • Provides protection from low-hazardous particles and liquids
  • Strong and abrasion-resistant
  • Suitable for multiple wearings
  • Seamless front to provide more protection in the primary exposure areas
  • Zipper front with 1 in. (2.5 cm) flap
  • Elastic back, wrists, ankles, and attached hood
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached
Closure Type Front Zipper
Sterility Non-sterile
Resistance Particle, Liquid
Back Style Elastic
Certifications/Compliance NFPA 99
Model KleenGuard™ A30
For Use With (Application) Pharmaceutical Research, Aerospace, Maintenance, General Manufacturing, Food
Quantity 25/Cs.
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.