Learn More
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
- Feature bound seams
- Storm flap provides a seamless chest area
- Water-resistant zipper and wick-away lining for enhanced comfort
- Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
Specifications
Specifications
| Size | 3X-Large |
| Material | Polyethylene, Spunbond Polypropylene |
| Color | Yellow |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Closure Type | Front Zipper with Storm Flap |
| Seam Type | Bound |
| Resistance | Chemicals |
| Certifications/Compliance | ASTM F 903, NFPA 99 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.