Learn More
Ansell KleenGuard™ A70 Chemical Spray Coveralls
Catalog No. 19166494 Shop All Ansell Products
Change view
Click to view available options
Description:
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-166-494 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood 2X-Large
19-166-488 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots 2X-Large
19-166-487 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots X-Large
19-166-489 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots 3X-Large
19-166-490 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots 4X-Large
19-166-491 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood Medium
19-166-492 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood Large
19-166-493 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood X-Large
19-166-496 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood 4X-Large
19-166-495 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood 3X-Large
19-166-485 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots Medium
19-812-900 Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood 5X-Large
19-166-486 Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
This item is not returnable. View return policy
Catalog No. 19-166-494 Supplier Ansell Supplier No. 09815
Add to Cart
Add to Cart
This item is not returnable. View return policy
Reflex™ design offers 12% more chest room and 6% longer body length
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
  • Feature bound seams
  • Storm flap provides a seamless chest area
  • Water-resistant zipper and wick-away lining for enhanced comfort
  • Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.
TRUSTED_SUSTAINABILITY
Specifications
Size 2X-Large
Material Polyethylene, Spunbond Polypropylene
Color Yellow
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Resistance Chemicals
Certifications/Compliance ASTM F 903, NFPA 99
Description Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood
Thickness 1.5 mil
For Use With (Application) Hazardous waste remediation, environmental cleanup, petrochemical refining, Acid/Caustic handling, biohazard cleanup, tank cleaning, pharmaceutical manufacturing