Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ansell KleenGuard™ A70 Chemical Spray Coveralls
Click to view available options
Description:
Zipper Front, Storm Flap, Elastic Wrists, Ankles and Hood
Zipper Front, Storm Flap, Elastic Wrists, Ankles, Hood and Boots
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
Coveralls provide protection for hazardous waste remediation, environmental cleanup, petrochemical and oil refining, acid and caustic handling, biohazard cleanup, tank cleaning and pharmaceutical manufacturing.
- Feature bound seams
- Storm flap provides a seamless chest area
- Water-resistant zipper and wick-away lining for enhanced comfort
- Yellow
Compliance
Meet ASTM™ F 903 penetration test standards.

Specifications
Specifications
Size | Medium |
Material | Polyethylene, Spunbond Polypropylene |
Color | Yellow |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Front Zipper with Storm Flap |
Seam Type | Bound |
Resistance | Chemicals |
Certifications/Compliance | ASTM F 903, NFPA 99 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction