Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G10 Arctic Blue Nitrile Gloves

Catalog No. 19021002 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-021-002 Medium
19-021-000 X-Small
19-021-001 Small
19-021-003 Large
19-021-004 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-021-002 Supplier KleenGuard™ Supplier No. 90097
This item is not returnable. View return policy

KleenGuard™ G10 gloves provide durability and strength for light-duty tasks. Ideal for industrial, manufacturing and food processing applications.

  • Powder free, nitrile
  • Textured fingertips provide enhanced grip and dexterity
  • Ambidextrous
  • Beaded cuff
  • Puncture-resistant formulation delivers barrier protection against chemical splash, dirt, grease and oil
  • Ideal for cleaning and maintenance, manufacturing, food processing, electronic assembly and painting applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA 21 CFR 170-199, CFIA approved
Applications Industrial
AQL 4
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium
Thickness (English) 2 mil
Thickness (Metric) 0.05 mm
Model 90097
Length (English) 9.5 in.
Length (Metric) 24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.