Description
- Designed with ergonomic features for long-term performance and all-day comfort
- Soft-touch temples naturally contour to the head for a secure and comfortable fit
- Cut-to-shape distinctive and stylish one-piece lens provides front and side protection
- Vents on temples maximize airflow and minimize fogging
- Flexible nosepiece conforms to various nose shapes; provides protection by minimizing gap between face and lens
Compliance
Meets ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
| Lens Tint | Smoke |
| Frame Color | Black |
| Lens Material | Polycarbonate |