Learn More
Ansell Smith & Wesson™ Caliber™ Safety Eyewear
Catalog No. 19706019 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Brown
Indoor/Outdoor
Smoke
Frame Color:
Black
Brown
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color
19-706-019 Indoor/Outdoor Black
19-706-020 Brown Brown
19-706-018 Smoke Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-706-019 Supplier Ansell Supplier No. 23008
This item is not returnable. View return policy
Precision, cut-to-shape spherical lens provides increased peripheral and distortion-free protection
  • Ergonomic, rubber nose pads with built-in airflow holes adjust to fit any nose shape
  • Scratch-resistant polycarbonate lens with antifog coating
  • Hinge stops prevent lens damage
  • Contoured, soft-touch rubber temples
  • Seven-base curve spherical lens
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3 standards.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1-2010 and CSA Z94.3 standards.
Lens Tint Indoor/Outdoor
Frame Color Black
Lens Material Polycarbonate