Description
Ergonomically dimensioned three-point fit for comfort
- Built-in hinge stops help to eliminate scratches
- Self-adjusting ergonomic rubber nose pad with rubber fingers
- Mini-Magnum™ designed to fit small faces
- Provides 99.9% UV protection
Compliance
Meet ANSI™ Z87.1-2010 standards.
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z87.1-2003 standards. |
| Lens Tint | GIO |
| Frame Color | Metallic Gray |
| Size | Full-sized |