Learn More
Ansell Smith & Wesson™ Phantom Safety Eyewear
Frameless design with lens, hinge, and nose pad post molded as one unit
Supplier:  Ansell 19838
 View more versions of this product
Catalog No. 19-130-110
This item is not returnable. View return policy
Description
Description
  • Seven-base spherical lens
  • Ribbed, non-slip rubber nose pad
  • Straight-back bayonet design temples eliminate mastoid bone pressure
  • UV protection
Compliance
Meets ANSI Z87.1 - 2010 High Velocity Industrial Standards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Anti-fog
Clear anti-fog lens
Straight back bayonet design
Exceeds ANSI Z87.1 - 2010 High Velocity Industrial Standards
Pewter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.