Learn More
Ansell Smith & Wesson™ Phantom Safety Eyewear
GREENER_CHOICE
Frameless design with lens, hinge, and nose pad post molded as one unit
$27.50
Specifications
Certifications/Compliance Exceeds ANSI Z87.1 - 2010 High Velocity Industrial Standards
Temple Style Straight back bayonet design
View More Specs
Products 3
Catalog Number Mfr. No. Lens Tint Frame Color Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Frame Color Price Quantity & Availability  
19-130-0904
GREENER_CHOICE
View Documents
Ansell
19853
Smoke Tint Lens Black
Each for $27.50
Pack of 12 EA for $308.00
Only null left
Add to Cart
19-130-0905
View Documents
Ansell
19839
Clear lens Pewter N/A N/A N/A
19-130-110
View Documents
Ansell
19838
Clear anti-fog lens Pewter N/A N/A N/A
Description
Description
  • Seven-base spherical lens
  • Ribbed, non-slip rubber nose pad
  • Straight-back bayonet design temples eliminate mastoid bone pressure
  • UV protection
Compliance
Meets ANSI Z87.1 - 2010 High Velocity Industrial Standards
Specifications
Specifications
Exceeds ANSI Z87.1 - 2010 High Velocity Industrial Standards
Straight back bayonet design
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.