Learn More
Ansell Visio™ Spectacle Lens
Catalog No. 18892182 Shop All Ansell Products
Change view
Click to view available options
XXX-Large
Catalog No.
18-892-182 XXX-Large
1 options
Catalog No. 18-892-182 Supplier Ansell Supplier No. 14472
Visio™ Spectacle Lens feature a sleek design with three-point extendable temples, providing a customized fit for enhanced comfort
  • Unique dual-done frame for added style
  • Lenses are hard coated for scratch resistance and longer life
  • Polycarbonate lenses that provide 99.9% UVA/UVB/UVC protection
  • Meet the ANSI Z87.1+ standards for impact and are TAA compliant.
  • Stylish dual-tone frame
TRUSTED_SUSTAINABILITY
Specifications
Description Spectacle Lens
Lens Tint Smoke
No. per Case 12/Cs.