Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ Scott™ Essential High Capacity Hard Roll Towels
Shop All Kimberly-Clark Professional Products
Click to view available options
For Use With (Equipment):
Kimberly-Clark Professional Dispensers
Universal Hard Roll Towel Dispensers
Description
- Scott Essential Hard Roll Paper Towel Rolls are 950 feet per roll, 9 rolls per case for a total of 5,700 feet
- High capacity: reduce maintenance time, run-outs and complaints with long-lasting bulk paper towels (as compared to a folded towel system)
- Compatible with select Kimberly-Clark Professional hard roll paper towel dispensers, designed to hold 1.75 inch cores (works with both manual and electronic dispensers)
- Highly absorbent commercial paper towels let you and your bathroom guests use less and waste less
- Paper towels come in space-saving cases for easy storage and handling at your business
Specifications
Specifications
Product Type | Hard Roll Towel |
Certifications/Compliance | EPA |
Color | White |
For Use With (Equipment) | Kimberly-Clark Professional Dispensers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction