Learn More
Ansell Fluidshield™ PFR95 N95 Particulate Filter Respirators and Surgical Masks
Catalog No. 189992454 Shop All Ansell Products
Change view
Click to view available options
Size:
Regular
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-2454 Small
18-999-2453 Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-2454 Supplier Ansell Supplier No. 46867
Fluidshield particulate filter respirator (PFR) and surgical mask with safety seal film meets NIOSH Approved standards.
  • Pouch-style with polyurethane
  • Headbands
  • Orange
  • Available in small and regular size
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color Orange
Design Type Pouch/Duckbill
Disposable Single-use
Filter Type Surgical N95
Product Type Healthcare Particulate Respirator and Surgical Mask
Size Small
With Exhalation Valve No
Filter Efficiency 95%
Model 46867
Quantity 1 Ea.
Resistant To Fluid
Strap Attachment Type Headbands