Learn More
Ansell Fluidshield™ PFR95 N95 Particulate Filter Respirators and Surgical Masks
Fluidshield particulate filter respirator (PFR) and surgical mask with safety seal film meets NIOSH Approved standards.
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color Orange
Design Type Pouch/Duckbill
Disposable Single-use
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-999-2454
View Documents
Ansell
46867
Small N/A N/A N/A
18-999-2453
View Documents
Ansell
46767
Regular N/A N/A N/A
Description
Description
  • Pouch-style with polyurethane
  • Headbands
  • Orange
  • Available in small and regular size
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Specifications
Specifications
N - Not Resistant to Oil
Orange
Single-use
Healthcare Particulate Respirator and Surgical Mask
95%
Fluid
NIOSH 42 CFR 84, N95
Pouch/Duckbill
Surgical N95
No
1 Ea.
Headbands
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.