Description
- Stylish
- Durable
- Sporty, flexible, lightweight design
- Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage
- Wraparound frame has a base curve of 8.25 that extends the range of peripheral protection
- Soft touch temples for comfort
- Each pair comes with a neck cord
- Manufactured to comply with the ANSI / ISEA Z87.1+ standard for personal eye protection and are CSA certified
- Comfortable and functional for construction, manufacturing, gun shooting / shooting range, and a host of other industries
Specifications
Specifications
| Lens Coating | Uncoated |
| Lens Tint | Smoke |
| Product Type | V30 NEMESIS Protective Spectacle |
| Frame Color | Black |
| Includes | Free neck cord |
| No. per Case | 12/Cs. |
| Temple Style | Soft touch |