Description
- Lightweight and stylish with single lens wraparound protection designed for use at work or home
- Flex-point temples help provide comfort and stability for use for long stretches of time
- Ideal for industrial manufacturin
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Hard coated lenses for scratch resistance and longer life
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Amber |
| Frame Color | Black |