Learn More
Ansell V40 HellRaiser™ Safety Glasses
V40 HellRaiser™ Safety Glasses feature a high-performance, durable polycarbonate lenses, which provide 99.9% UV protection (UVA/UVB/UVC)
Supplier: Ansell 20544
This item is not returnable.
View return policy
Description
- Lightweight and stylish with single lens wraparound protection designed for use at work or home
- Flex-point temples help provide comfort and stability for use for long stretches of time
- Ideal for industrial manufacturin
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Hard coated lenses for scratch resistance and longer life
Specifications
Scratch-resistant | |
IR Filter Shade 3.0 | |
Black |