Learn More
Ansell V40 HellRaiser™ Safety Glasses
V40 HellRaiser™ Safety Glasses feature a high-performance, durable polycarbonate lenses, which provide 99.9% UV protection (UVA/UVB/UVC)
$11.09 - $287.64
Catalog Number | Mfr. No. | Lens Tint | Lens Coating | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Lens Coating | Price | Quantity & Availability | ||||
19-169-871
|
Ansell
28615 |
Clear | Anti-fog, Scratch-resistant |
Case of 12 for $287.64
|
|
||||
19-800-002
![]() |
Ansell
25716 |
Indoor/Outdoor | Scratch-resistant |
Pair for $24.58
Case of 12 PR for $295.02
|
|
||||
19-800-000
![]() |
Ansell
20539 |
Clear | Scratch-resistant |
Pair for $20.83
Case of 12 PR for $250.01
|
|
||||
19-800-001
![]() |
Ansell
25714 |
Smoke | Scratch-resistant |
Pair for $11.09
Case of 12 PR for $133.11
|
|
||||
19-800-005
|
Ansell
20543 |
Blue Mirror | Scratch-resistant |
Pair for $16.09
Case of 12 PR for $193.03
|
|
||||
19-800-007
|
Ansell
20545 |
IR Filter Shade 5.0 | Scratch-resistant |
Pair for $25.47
Case of 12 PR for $305.61
|
|
||||
19-800-003
|
Ansell
20541 |
Amber | Scratch-resistant | N/A | N/A | N/A | |||
19-800-004
|
Ansell
20542 |
Blue | Scratch-resistant | N/A | N/A | N/A | |||
19-800-006
|
Ansell
20544 |
IR Filter Shade 3.0 | Scratch-resistant | N/A | N/A | N/A | |||
Description
- Lightweight and stylish with single lens wraparound protection designed for use at work or home
- Flex-point temples help provide comfort and stability for use for long stretches of time
- Ideal for industrial manufacturin
- Polycarbonate lenses that provide 99.9% UVA/ UVB/UVC protection to help prevent eye damage like cataracts, retinal damage, and other conditions that can cause temporary vision loss
- Hard coated lenses for scratch resistance and longer life
Specifications
Black |