Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Black with Cone-Shaped Liner Phenolic Caps/Closures
Shop All DWK Life Sciences Products

Click to view available options
Closure Size:
13-425
15-425
18-400
22-400
Description
- Phenolic
- Cone-shaped liner for two-part seal
- Superior torque retention

Specifications
Specifications
Material (Liner) | Polyethylene |
Color | Black |
Material | Phenolic |
For Use With (Application) | Excellent sample storage and re-sealing properties |
Closure Size | 13-425 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction