Learn More
DWK Life Sciences Kimble™ Brand Rotary Evaporator: Bottom Vapor Tube Traps
Accessory for rotary evaporator trap
$287.00 - $346.00
Specifications
Certifications/Compliance | USP Type I and ASTM E438, Class A |
---|---|
Material | Borosilicate Glass |
Product Type | Bottom Vapor Tube Trap |
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Standard Taper Joint | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (English) | Capacity (Metric) | Standard Taper Joint | For Use With (Application) | Price | Quantity & Availability | ||
K5702600114
![]() |
DWK Life Sciences
5702600114 |
3.4 oz. | 100 mL | Upper: 24/40, Lower: 14/20 | Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping |
Case for $295.00
|
|
||
K5702602514
![]() |
DWK Life Sciences
5702602514 |
8.45 oz. | 250 mL | Upper: 24/40, Lower: 14/20 | Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping |
Case for $287.00
|
|
||
K5702600124
![]() |
DWK Life Sciences
5702600124 |
3.4 oz. | 100 mL | Upper: 24/40, Lower: 24/40 | Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping |
Case for $293.00
|
|
||
K5702652514
![]() |
DWK Life Sciences
5702652514 |
8.45 oz. | 250 mL | Upper: 24/40, Lower: 14/20 | Used when foam is likely to be created and drawn into the condenser of rotary evaporator |
Case for $342.00
|
|
||
K5702605024
![]() |
DWK Life Sciences
5702605024 |
16.9 oz. | 500 mL | Upper: 24/40, Lower: 24/40 | Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping |
Case for $310.00
|
|
||
K5702652524
![]() |
DWK Life Sciences
5702652524 |
8.45 oz. | 250 mL | Upper: 24/40, Lower: 24/40 | Used when foam is likely to be created and drawn into the condenser of rotary evaporator |
Case for $346.00
|
|
||
K5702602524
![]() |
DWK Life Sciences
5702602524 |
8.45 oz. | 250 mL | Upper: 24/40, Lower: 24/40 | Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping |
Case for $303.00
|
|
||
K5702655024
![]() |
DWK Life Sciences
5702655024 |
16.9 oz. | 500 mL | Upper: 24/40, Lower: 24/40 | Used when foam is likely to be created and drawn into the condenser of rotary evaporator |
Case for $337.00
|
|
||
Description
- Square Joint Bead
Specifications
USP Type I and ASTM E438, Class A | |
Borosilicate Glass | |
Bottom Vapor Tube Trap |