Learn More
DWK Life Sciences™ Kimble™ Kontes™ Claisen Distillation Adapter
SDP
Provides dual entry into a variety of flasks
$197.90
Specifications
For Use With (Application) For reflux with addition-type reactions, for determining the fluoride ion in water samples
Type Claisen Distillation Adapter
Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
View More Specs
Products 1
Catalog Number Mfr. No. Standard Taper Joint Height (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Standard Taper Joint Height (English) Width (English) Price Quantity & Availability  
K161500-2440
SDP
View Documents
DWK Life Sciences™
1615002440
24/40 6.1 in. 3.6 in.
Case of 1 for $197.90
Only null left
Add to Cart
Description
Description
  • For determining the fluoride ion in water samples
  • Fluoride ion is isolated in the distillation
Specifications
Specifications
For reflux with addition-type reactions, for determining the fluoride ion in water samples
Claisen Distillation Adapter
USP Type I, ASTM E438, Type I, Class A
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.