Learn More
DWK Life Sciences Kimble™ Kontes™ Cyanide Distillation Apparatus with Sulfide Scrubber
Click to view available options
:
Cyanide distillation apparatus
Description
- Samples react with acid to convert cyanide to HCN
- The first bubbler acts as a scrubber to remove sulfides
- Cyanide is absorbed in the second bubbler
- Design features compact Standard Taper 19/38 joints
- Capacity 1000mL
- Feature Upper Standard Taper
- Standard Taper joints 19/38
- Case of 1
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
Certifications/Compliance | USP Type I, ASTM E438, Type I, Class A |
For Use With (Application) | Quantitative analysis of various industrial wastewaters, saline, and reagent-grade water |
Type | Cyanide Distillation Apparatus with Sulfide Scrubber |