Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences™ Kimble™ Kontes™ MIDI-VAP 4000 Component Replacement Parts
SDP

Catalog No. p-4244400 Shop All DWK Life Sciences Products
Click to view available options
Type:
Ammonia Phenol Glassware 1 Set
Ammonia Phenol Glassware 5 Sets
Closed Top Screw Thread Cap
Cyanide Distillation
Cyanide Glassware Set
Distilling Head
Long Stem Outlet Tube
Manifold Heater
Open Top Screw Thread Cap
Quick Disconnect Fittings, Female
Quick Disconnect Fittings, Male
Reaction Tube
Reaction/Absorption Tube
Receiver Tube
Sealing Ring
Short Stem Outlet Tube
Tubing Kit
For Use With (Equipment):
For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
MIDI-VAP 4000
MIDI-VAP 4000 Ammonia System
MIDI-VAP 4000 Ammonia/Phenol System
MIDI-VAP 4000 Cyanide System
MIDI-VAP 4000 Phenol System

Catalog No. K4104800014

Supplier: DWK Life Sciences™ 4104800014

Only null left
Add to Cart
Add to Cart

Component parts for both ammonia and phenol distillation system

Component parts for both ammonia and phenol distillation system

  • For use with MIDI-VAP 4000 Distillation Apparatus
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Distillation systems
Type Sealing Ring
Material PTFE/Silicone Liner
For Use With (Equipment) For GL-14 cap, MIDI-VAP 4000 Ammonia/Phenol System
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.